Assay Details
Mature miRNA Sequence: | UCCCUGUCCUCCAGGAGCUCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Pig | ssc-miR-339 | MIMAT0025372 |
| Pteropus alecto | pal-miR-339a-5p | MIMAT0040284 |
Stem-loop Details
Pig
Pteropus alecto
More Information
Related Products
miRBase Alias:
Human: hsa-miR-339 (v9.2)
Mouse: mmu-miR-339 (v9.2)
Rat: rno-miR-339 (v9.2)
Mouse: mmu-miR-339 (v9.2)
Rat: rno-miR-339 (v9.2)

Genomic Map