Assay Details
Gene Family ID: | MIPF0000133, mir-449 |
Mature miRNA Sequence: | UGGCAGUGUAUUGUUAGCUGGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-449a | MIMAT0001541 | |
| Mouse | mmu-miR-449a-5p | MIMAT0001542 | |
| Rat | rno-miR-449a-5p | MIMAT0001543 | |
| Bovine | bta-miR-449a | MIMAT0009320 | |
| Callithrix jacchus | cja-miR-449a | MIMAT0039495 | |
| Cricetulus griseus | cgr-miR-449a | MIMAT0041140 | |
| View More Rows | |||
| Dasypus novemcinctus | dno-miR-449a-5p | MIMAT0047892 | |
| Dog | cfa-miR-449a | MIMAT0001544 | |
| Guinea pig | cpo-miR-449a-5p | MIMAT0047257 | |
| Horse | eca-miR-449a | MIMAT0013102 | |
| Monodelphis domestica | mdo-miR-449a-5p | MIMAT0004140 | |
| Pongo pygmaeus | ppy-miR-449a | MIMAT0015891 | |
| Pteropus alecto | pal-miR-449a-5p | MIMAT0039939 | |
| Rabbit | ocu-miR-449a-5p | MIMAT0048453 | |
| Rhesus monkey | mml-miR-449a-5p | MIMAT0006330 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Cricetulus griseus
Dasypus novemcinctus
Dog
Guinea pig
Horse
Monodelphis domestica
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Human Pool AMegaplex™ RT Primers, Rodent Pool A
Related Products
miRBase Alias:
Human: hsa-miR-449 (v9.2)
Mouse: mmu-miR-449 (v9.2)
Mouse: mmu-miR-449a (v17)
Rat: rno-miR-449a (v18)
Rat: rno-miR-449 (v9.2)
Citrobacter farmeri: cfa-miR-449 (v20)
Macaca mulatta: mml-miR-449a (v19)
Monodelphis domestica: mdo-miR-449 (v19)
Mouse: mmu-miR-449 (v9.2)
Mouse: mmu-miR-449a (v17)
Rat: rno-miR-449a (v18)
Rat: rno-miR-449 (v9.2)
Citrobacter farmeri: cfa-miR-449 (v20)
Macaca mulatta: mml-miR-449a (v19)
Monodelphis domestica: mdo-miR-449 (v19)

Genomic Map