Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | AAUAUUAUACAGUCAACCUCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-656-3p | MIMAT0003332 | |
| Bovine | bta-miR-656 | MIMAT0009361 | |
| Dasypus novemcinctus | dno-miR-656-3p | MIMAT0047993 | |
| Goat | chi-miR-656 | MIMAT0036283 | |
| Guinea pig | cpo-miR-656-3p | MIMAT0047364 | |
| Horse | eca-miR-656 | MIMAT0013172 | |
| View More Rows | |||
| Pan troglodytes | ptr-miR-656 | MIMAT0008305 | |
| Pongo pygmaeus | ppy-miR-656 | MIMAT0016090 | |
| Pteropus alecto | pal-miR-656-3p | MIMAT0040368 | |
| Rabbit | ocu-miR-656-3p | MIMAT0048550 | |
| Rhesus monkey | mml-miR-656-3p | MIMAT0006497 | |
| Tupaia chinensis | tch-miR-656 | MIMAT0036556 | |
Stem-loop Details
Human
Bovine
Dasypus novemcinctus
Goat
Guinea pig
Horse
Pan troglodytes
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-656 (v19)
Macaca mulatta: mml-miR-656 (v19)
Macaca mulatta: mml-miR-656 (v19)

Genomic Map