Assay Details
Gene Family ID: | MIPF0000073, mir-129 |
Mature miRNA Sequence: | AAGCCCUUACCCCAAAAAGUAU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-129-1-3p | MIMAT0004548 | |
| Mouse | mmu-miR-129-1-3p | MIMAT0016994 | |
| Alligator mississippiensis | ami-miR-129a-3p | MIMAT0038137 | |
| Dasypus novemcinctus | dno-miR-129-3p | MIMAT0047625 | |
| Guinea pig | cpo-miR-129-3p | MIMAT0046978 | |
| Horse | eca-miR-129a-3p | MIMAT0012933 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-129-1-3p | MIMAT0015752 | |
| Pteropus alecto | pal-miR-129b-3p | MIMAT0039873 | |
| Rabbit | ocu-miR-129-3p | MIMAT0048215 | |
| Rhesus monkey | mml-miR-129-1-3p | MIMAT0006186 | |
| Salmo salar | ssa-miR-129-3-3p | MIMAT0032320 | |
| Zebrafish | dre-miR-129-3-3p | MIMAT0048649 | |
Stem-loop Details
Human
Mouse
Alligator mississippiensis
Dasypus novemcinctus
Guinea pig
Horse
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey
Salmo salar
Zebrafish
More Information
Related Products
miRBase Alias:
Human: hsa-miR-129* (v17)
Macaca mulatta: mml-miR-129-3p (v19)
Pongo pygmaeus: ppy-miR-129-1* (v18)
Macaca mulatta: mml-miR-129-3p (v19)
Pongo pygmaeus: ppy-miR-129-1* (v18)

Genomic Map