Assay Details
Gene Family ID: | MIPF0000082, mir-193 |
Mature miRNA Sequence: | AACUGGCCCUCAAAGUCCCGCU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-193b-3p | MIMAT0002819 | |
| Callithrix jacchus | cja-miR-193 | MIMAT0039349 | |
| Nomascus leucogenys | nle-miR-193b | MIMAT0049412 | |
| Pan troglodytes | ptr-miR-193b | MIMAT0008059 | |
| Papio hamadryas | pha-miR-193b | MIMAT0049567 | |
| Pongo pygmaeus | ppy-miR-193b | MIMAT0015789 | |
| View More Rows | |||
| Rhesus monkey | mml-miR-193b-3p | MIMAT0006227 | |
Stem-loop Details
Human
Callithrix jacchus
Nomascus leucogenys
Pan troglodytes
Papio hamadryas
Pongo pygmaeus
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-193b (v17)
Macaca mulatta: mml-miR-193b (v19)
Macaca mulatta: mml-miR-193b (v19)

Genomic Map