Assay Details
Gene Family ID: | MIPF0000009, mir-29 |
Mature miRNA Sequence: | ACUGAUUUCUUUUGGUGUUCAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-29a-5p | MIMAT0004503 |
| Mouse | mmu-miR-29a-5p | MIMAT0004631 |
| Rat | rno-miR-29a-5p | MIMAT0004718 |
| Ornithorhynchus anatinus | oan-miR-29a-1-5p | MIMAT0006806 |
| Pig | ssc-miR-29a-5p | MIMAT0037364 |
| Pteropus alecto | pal-miR-29a-5p | MIMAT0039801 |
Stem-loop Details
Human
Mouse
Rat
Ornithorhynchus anatinus
Pig
Pteropus alecto
More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
miRBase Alias:
Human: hsa-miR-29a* (v17)
Mouse: mmu-miR-29a* (v17)
Rat: rno-miR-29a* (v18)
Ochrobactrum anthropi: oan-miR-29a-1* (v18)
Mouse: mmu-miR-29a* (v17)
Rat: rno-miR-29a* (v18)
Ochrobactrum anthropi: oan-miR-29a-1* (v18)

Genomic Map