Assay Details
Gene Family ID: | MIPF0000038, mir-1 |
Mature miRNA Sequence: | UACAUACUUCUUUACAUUCCA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-1b-5p | MIMAT0005835 |
| Callithrix jacchus | cja-miR-1b-5p | MIMAT0039616 |
Stem-loop Details
Mouse
Callithrix jacchus
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-1-2-as (v15)
Mouse: mmu-miR-1-2-as-5p (v16)
Mouse: mmu-miR-1-2-as-5p (v16)

Genomic Map