Assay Details
Gene Family ID: | MIPF0000022, mir-7 |
Mature miRNA Sequence: | UGGAAGACUAGUGAUUUUGUUGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-7-5p | MIMAT0000252 | |
| Acyrthosiphon pisum | api-miR-7 | MIMAT0014734 | |
| Bactrocera dorsalis | bdo-miR-7 | MIMAT0045389 | |
| Bovine | bta-miR-7 | MIMAT0003843 | |
| Callithrix jacchus | cja-miR-7 | MIMAT0039489 | |
| Columba livia | cli-miR-7a-5p | MIMAT0038396 | |
| View More Rows | |||
| Cyprinus carpio | ccr-miR-7a | MIMAT0026319 | |
| Dasypus novemcinctus | dno-miR-7-5p | MIMAT0047491 | |
| Dinoponera quadriceps | dqu-miR-7-5p | MIMAT0045643 | |
| Drosophila virilis | dvi-miR-7-5p | MIMAT0008931 | |
| Gadus morhua | gmo-miR-7a-5p | MIMAT0044123 | |
| Goat | chi-miR-7-5p | MIMAT0036288 | |
| Guinea pig | cpo-miR-7-5p | MIMAT0046839 | |
| Lottia gigantea | lgi-miR-7 | MIMAT0009560 | |
| Manduca sexta | mse-miR-7 | MIMAT0024426 | |
| Metriaclima zebra | mze-miR-7a | MIMAT0042370 | |
| Ophiophagus hannah | oha-miR-7-5p | MIMAT0036945 | |
| Pig | ssc-miR-7-5p | MIMAT0002141 | |
| Polistes canadensis | pca-miR-7-5p | MIMAT0045834 | |
| Pteropus alecto | pal-miR-7b-5p | MIMAT0039917 | |
| Python bivittatus | pbv-miR-7-5p | MIMAT0038817 | |
| Rabbit | ocu-miR-7a-5p | MIMAT0048093 | |
| Tetranychus urticae | tur-miR-7-5p | MIMAT0023036 | |
| Triops cancriformis | tcf-miR-7 | MIMAT0041346 | |
| Tupaia chinensis | tch-miR-7-5p | MIMAT0036465 | |
Stem-loop Details
Human
| Stem-loop ID | hsa-mir-7-1 |
| Stem-loop Accession # | MI0000263 |
| Stem-loop Sequence | UUGGAUGUUGGCCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAUAUGGCACAGGCCAUGCCUCUACAG |
| Chromosome Location | Chr. 9 - 83969748 - 83969857 [-] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000252
miRBase ID: hsa-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: Chr. 19 - 4770670 - 4770779 [+] on Build GRCh38 |
| Stem-loop ID | hsa-mir-7-2 |
| Stem-loop Accession # | MI0000264 |
| Stem-loop Sequence | CUGGAUACAGAGUGGACCGGCUGGCCCCAUCUGGAAGACUAGUGAUUUUGUUGUUGUCUUACUGCGCUCAACAACAAAUCCCAGUCUACCUAAUGGUGCCAGCCAUCGCA |
| Chromosome Location | Chr. 15 - 88611825 - 88611934 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000252
miRBase ID: hsa-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: Chr. 19 - 4770670 - 4770779 [+] on Build GRCh38 |
| Stem-loop ID | hsa-mir-7-3 |
| Stem-loop Accession # | MI0000265 |
| Stem-loop Sequence | AGAUUAGAGUGGCUGUGGUCUAGUGCUGUGUGGAAGACUAGUGAUUUUGUUGUUCUGAUGUACUACGACAACAAGUCACAGCCGGCCUCAUAGCGCAGACUCCCUUCGAC |
| Chromosome Location | Chr. 19 - 4770670 - 4770779 [+] on Build GRCh38 |
| Mature MicroRNA |
miRBase Accession #: MIMAT0000252
miRBase ID: hsa-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: Chr. 19 - 4770670 - 4770779 [+] on Build GRCh38 |
| Stem-loop ID | api-mir-7 |
| Stem-loop Accession # | MI0013936 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUAAAAAAAAAGUAUUUUCCUAACAAGAAGUCUCUAGUCUUCUUAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0014734
miRBase ID: api-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | bdo-mir-7 |
| Stem-loop Accession # | MI0037266 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUGUUUCACGAGUAACAAUAAAUCCCUUGUCUUCUUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0045389
miRBase ID: bdo-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | bta-mir-7-1 |
| Stem-loop Accession # | MI0010471 |
| Stem-loop Sequence | GUGGAGCAGCCAGCCCCAUCUGGAAGACUAGUGAUUUUGUUGUUGUCUCCUGCGCUCAACAACAAGUCCCAGUCUGCCGCAUGGUGCUGGCCACCGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003843
miRBase ID: bta-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | bta-mir-7-2 |
| Stem-loop Accession # | MI0010462 |
| Stem-loop Sequence | UGGAUGUUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUGAAUCGACAACAAAUCACAGUCUGCCAUAUGGCACAGGCCAUGCCUCUACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003843
miRBase ID: bta-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | bta-mir-7-3 |
| Stem-loop Accession # | MI0005056 |
| Stem-loop Sequence | AGACUGGAGCGGUUGUGGUCUAGUGCUGUGUGGAAGACUAGUGAUUUUGUUGUUCUGAUGUACUAUGACAACAAGUCACAGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0003843
miRBase ID: bta-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cja-mir-7 |
| Stem-loop Accession # | MI0032013 |
| Stem-loop Sequence | GGAUAUUGGCCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUUGACAACAAAUCACAGUCUGCCAUAUGGCACAGGCCAUGCC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039489
miRBase ID: cja-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cli-mir-7a-1 |
| Stem-loop Accession # | MI0029896 |
| Stem-loop Sequence | GUUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUUGACAACAAAUCGCAGUCUGCCAUAUAGCACAGACUGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038396
miRBase ID: cli-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cli-mir-7a-2 |
| Stem-loop Accession # | MI0029897 |
| Stem-loop Sequence | GGUGCCCAGCUCCCUCUGGAAGACUAGUGAUUUUGUUGUUGUAUGGCUCAUCCCACCACAACAAGUCACAGUCUGCCCUAGGGUGCGUGGCCUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038396
miRBase ID: cli-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cli-mir-7a-3 |
| Stem-loop Accession # | MI0029898 |
| Stem-loop Sequence | GGUGGUCUGGCUCUGCGUGGAAGACUAGUGAUUUUGUUGUUCUGAUUUAUAAAGGUGACAACAAAUCAUAGCCUGCCACCCAGCCCAGACCUGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038396
miRBase ID: cli-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | ccr-mir-7a |
| Stem-loop Accession # | MI0023416 |
| Stem-loop Sequence | AUUUUGGGCACCAUGGAAGACUAGUGAUUUUGUUGUUGUGAAGUCACCGGUGCUGACAACAAGUCACAGUCUGCCUCAGUGACCAGAAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0026319
miRBase ID: ccr-miR-7a Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | dno-mir-7-1 |
| Stem-loop Accession # | MI0038897 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUUGACAACAAAUCACAGUCUGCCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047491
miRBase ID: dno-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | dno-mir-7-2 |
| Stem-loop Accession # | MI0038898 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUGUCUCACUGUGCGCUCAACAACAAGUCCCAGUCUGCCGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047491
miRBase ID: dno-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | dno-mir-7-3 |
| Stem-loop Accession # | MI0038899 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUCUGAUGUACUAUGACAACAAAUCACAGCCGGCCUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0047491
miRBase ID: dno-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | dqu-mir-7 |
| Stem-loop Accession # | MI0037471 |
| Stem-loop Sequence | CCGUUGUAUGGAAGACUAGUGAUUUUGUUGUUCUACUUUCGAUGUAACAAGGAAUCACUAAUCAUCCUACAAAGGCGCUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0045643
miRBase ID: dqu-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | dvi-mir-7 |
| Stem-loop Accession # | MI0009497 |
| Stem-loop Sequence | AAUGCUUUCCGUAUGGAAGACUAGUGAUUUUGUUGUUUGGUCUCUGCUAAUAACAAUAAAUCCCUUGUCUUCUUAUGGCGCGCAUU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0008931
miRBase ID: dvi-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-7a-1 |
| Stem-loop Accession # | MI0036057 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGUUAGAUGAACCGACAACAAAUCACAGUCUGCCAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044123
miRBase ID: gmo-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-7a-2 |
| Stem-loop Accession # | MI0036118 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUCUGUAAAUGAACCGACAACAAGUCAUUGUCUUCCUUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044123
miRBase ID: gmo-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | gmo-mir-7a-3 |
| Stem-loop Accession # | MI0036167 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUGUUAAAUAUUGACUUUCAUGACAACAAGUCUCUGUCUCCCUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0044123
miRBase ID: gmo-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | chi-mir-7 |
| Stem-loop Accession # | MI0030831 |
| Stem-loop Sequence | UUAUACAGAGCCUGUAGAAAAUGUAGAAGAUUCAGUGGAUGUUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUGAAUCGACAACAAAUCACAGUCUGCCAUAUGGCACAGGCCAUGCCUCUACAGGAC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036288
miRBase ID: chi-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-7-1 |
| Stem-loop Accession # | MI0038553 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046839
miRBase ID: cpo-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-7-2 |
| Stem-loop Accession # | MI0038554 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUGUCUCACUGUGCCCAACAACAAGUCCCAGUCUGCCGCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046839
miRBase ID: cpo-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | cpo-mir-7-3 |
| Stem-loop Accession # | MI0038555 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUCUGAUGUUGGAUGACAACAAGUCACAGCCGGCCACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0046839
miRBase ID: cpo-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | lgi-mir-7 |
| Stem-loop Accession # | MI0010103 |
| Stem-loop Sequence | AUAACCUAUCUACCCUGGUCUGGAAGACUAGUGAUUUUGUUGUUUUAAUUUCAGUAUUAACAAUAAAUCACAUUCUUCCCAGAUUGUGGUUGAUCUUAACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0009560
miRBase ID: lgi-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | mse-mir-7 |
| Stem-loop Accession # | MI0020975 |
| Stem-loop Sequence | CAUCGAGGAGCCGCGCGUUGUAUGGAAGACUAGUGAUUUUGUUGUUUUGAUUAAUUAACAAGGAAUCACUAAUCUGCCUACAAAGCGUCGGCAUGCGCUUCGGA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0024426
miRBase ID: mse-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | mze-mir-7a-1 |
| Stem-loop Accession # | MI0034157 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUGUAAAUUCAUGUUUUUGACAACAGAGUCACAGUCUACCUC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042370
miRBase ID: mze-miR-7a Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | mze-mir-7a-2 |
| Stem-loop Accession # | MI0034152 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGUUAGAGCAACCGACAACAAAUCACAGUCUGCCAAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042370
miRBase ID: mze-miR-7a Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | mze-mir-7a-3 |
| Stem-loop Accession # | MI0034193 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUGUUGUUUAAAAGGACAACAAAUCAUUGUCUCCCUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0042370
miRBase ID: mze-miR-7a Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | oha-mir-7-1 |
| Stem-loop Accession # | MI0031482 |
| Stem-loop Sequence | UCAGUUGGACGUUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUUACUAAAGUGACAACAAAUCGCAGUCUACCAUAUGGCACAGUCCAUGCCUCUACAGGACAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036945
miRBase ID: oha-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | oha-mir-7-2 |
| Stem-loop Accession # | MI0031483 |
| Stem-loop Sequence | CUUAUGGAAGACUAGUGAUUUUGUUGUUUCUGUAGCUCAUCUCAUGACAACAAGUCGCAGUCUGCCUUAUGGGGUCUGGCCCUGCCUCAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036945
miRBase ID: oha-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | oha-mir-7-3 |
| Stem-loop Accession # | MI0031484 |
| Stem-loop Sequence | AGACCUGGCCUGCUUCUGAUUGGAAGACUAGUGAUUUUGUUGUUGUGGUGUAGAAAAGCGACAACAAAUCACAGCCUACCAUCCAGCAGGGG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036945
miRBase ID: oha-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-7-1 |
| Stem-loop Accession # | MI0022122 |
| Stem-loop Sequence | CAGCCCCAUCUGGAAGACUAGUGAUUUUGUUGUUGUCUCACUGCGCUCAACAACAAGUCCCAGUCUGCCGCAUGGUGCCGGCCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002141
miRBase ID: ssc-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | ssc-mir-7-2 |
| Stem-loop Accession # | MI0002435 |
| Stem-loop Sequence | UGGAUGUUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAUAUGGCACAGGCCAUGCCUCUACAG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0002141
miRBase ID: ssc-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | pca-mir-7 |
| Stem-loop Accession # | MI0037570 |
| Stem-loop Sequence | AAAGAAAGAGCUCCGUUGUAUGGAAGACUAGUGAUUUUGUUGUUCUACUUUGGAUGUAACAAGGAAUCACUAAUCAUCCUACAAAGGGACUCGCAUAUCGUCGGCUACCUGGUG |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0045834
miRBase ID: pca-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | pal-mir-7b-1 |
| Stem-loop Accession # | MI0032399 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAU |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039917
miRBase ID: pal-miR-7b-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | pal-mir-7b-2 |
| Stem-loop Accession # | MI0032527 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUCUGAUGUACAAUGACAACAAGUCACAGCUUCCUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0039917
miRBase ID: pal-miR-7b-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-7-1 |
| Stem-loop Accession # | MI0030146 |
| Stem-loop Sequence | UUGGUCUAGUUCUGUGUGGAAGACUAGUGAUUUUGUUGUUUUUAGAUUACUAAAGUGACAACAAAUCGCAGUCUACCAUAUGGCACAGUCCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038817
miRBase ID: pbv-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-7-2 |
| Stem-loop Accession # | MI0030147 |
| Stem-loop Sequence | UGCCGGACUCCUUAUGGAAGACUAGUGAUUUUGUUGUUUCUGUAGCUCAUCUCAUGACAACAAGUCACAGUCUGCCUUCUGGAGUCUGGC |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038817
miRBase ID: pbv-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | pbv-mir-7-3 |
| Stem-loop Accession # | MI0030148 |
| Stem-loop Sequence | GGCCUGCUUCUGAUUGGAAGACUAGUGAUUUUGUUGUUCUGGUUUAGAAAGCUGACAACAAAUCACAGCCUACCAUCCAGCAGGGGCCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0038817
miRBase ID: pbv-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | ocu-mir-7a |
| Stem-loop Accession # | MI0039216 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0048093
miRBase ID: ocu-miR-7a-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | tur-mir-7 |
| Stem-loop Accession # | MI0019363 |
| Stem-loop Sequence | AUCUCCUUGUGGAAGACUAGUGAUUUUGUUGUUUGUCUGUUAGUAAUUAAACAAGAAAUCGCUAGCUUCCGAUGGAGAA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0023036
miRBase ID: tur-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | tcf-mir-7 |
| Stem-loop Accession # | MI0033192 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUCGAUAUAUGGGGAAACAAAAAGUCACUAGUUAUCCUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0041346
miRBase ID: tcf-miR-7 Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | tch-mir-7-1 |
| Stem-loop Accession # | MI0031124 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUCUGAUAUGUGAUGACAACAAGUCACAGCCGGCCUCA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036465
miRBase ID: tch-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | tch-mir-7-2 |
| Stem-loop Accession # | MI0031183 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUGUGUCACUAUACUCAACAACAAGUCCCAGUCUGCCACA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036465
miRBase ID: tch-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | tch-mir-7-3 |
| Stem-loop Accession # | MI0031202 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036465
miRBase ID: tch-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
| Stem-loop ID | tch-mir-7-4 |
| Stem-loop Accession # | MI0031288 |
| Stem-loop Sequence | UGGAAGACUAGUGAUUUUGUUGUUUUUAGAUAACUAAAUCGACAACAAAUCACAGUCUGCCAUA |
| Chromosome Location | - |
| Mature MicroRNA |
miRBase Accession #: MIMAT0036465
miRBase ID: tch-miR-7-5p Mature miRNA Sequence: UGGAAGACUAGUGAUUUUGUUGUU Chromosome Location: NA |
More Information
Related Products
miRBase Alias:
Staphylococcus schleiferi: ssc-miR-7 (v21)

Genomic Map