Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCACCAGCCUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila erecta | der-miR-311c-3p | MIMAT0008522 |
Stem-loop Details
Drosophila erecta
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | der-miR-311c |
|---|---|
| miRBase Accession Number: | MI0009062 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAUUGCACAUUCACCAGCCUGA |
| Species: | Drosophila erecta |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACAUUCACCAGCCUGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila erecta | der-miR-311c-3p | MIMAT0008522 |