Assay Details
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | GCACUGGGUGAAGUUUGUCUUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila persimilis | dpe-miR-309 | MIMAT0008760 |
| Drosophila pseudoobscura | dps-miR-309 | MIMAT0001259 |
Stem-loop Details
Drosophila persimilis
Drosophila pseudoobscura

Genomic Map