Assay Details
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UCACUGGGCAUAGUUUGUCGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Culex quinquefasciatus | cqu-miR-309 | MIMAT0014444 |
Stem-loop Details
Culex quinquefasciatus
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | cqu-miR-309 |
|---|---|
| miRBase Accession Number: | MI0013631 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACUGGGCAUAGUUUGUCGC |
| Species: | Culex quinquefasciatus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UCACUGGGCAUAGUUUGUCGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Culex quinquefasciatus | cqu-miR-309 | MIMAT0014444 |