Assay Details
Gene Family ID: | MIPF0001261, mir-3529 |
Mature miRNA Sequence: | AGGUAGACUGGGAUUUGUUGUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3529-5p | MIMAT0019828 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-3529 |
|---|---|
| miRBase Accession Number: | MI0017351 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | AGGUAGACUGGGAUUUGUUGUU |
| Species: | Human |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001261, mir-3529 |
Mature miRNA Sequence: | AGGUAGACUGGGAUUUGUUGUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3529-5p | MIMAT0019828 |