Assay Details
Gene Family ID: | MIPF0001711, mir-1545 |
Mature miRNA Sequence: | AGUGCCCUGGGAUAGAUAAGAGGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-1545c-5p | MIMAT0028752 |
Stem-loop Details
Monodelphis domestica

Genomic Map