Assay Details
Gene Family ID: | MIPF0001645, mir-1544 |
Mature miRNA Sequence: | ACUUUUCUAUCCCUGGUACUGGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Monodelphis domestica | mdo-miR-1544c-3p | MIMAT0028747 |
Stem-loop Details
Monodelphis domestica

Genomic Map