Assay Details
Gene Family ID: | MIPF0000101, MIR390 |
Mature miRNA Sequence: | CGCUAUCUAUCCUGAGCUCC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Aegilops tauschii | ata-miR390-3p | MIMAT0037141 |
| Rice | osa-miR390-3p | MIMAT0022888 |
Stem-loop Details
Aegilops tauschii
Rice

Genomic Map