Assay Details
Mature miRNA Sequence: | UUUGUACUUCAUCAGGUGCUCUG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tetranychus urticae | tur-miR-305-5p | MIMAT0023114 |
Stem-loop Details
Tetranychus urticae
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | tur-miR-305-5p |
|---|---|
| miRBase Accession Number: | MI0019409 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUUGUACUUCAUCAGGUGCUCUG |
| Species: | Tetranychus urticae |
| Product Type: | TaqMan™ MicroRNA Assay |
Mature miRNA Sequence: | UUUGUACUUCAUCAGGUGCUCUG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Tetranychus urticae | tur-miR-305-5p | MIMAT0023114 |