Assay Details
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | GUGAGAACUCUGAGGAUAAG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-454-5p | MIMAT0021962 |
Stem-loop Details
Anolis carolinensis
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | aca-miR-454-5p |
|---|---|
| miRBase Accession Number: | MI0018870 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GUGAGAACUCUGAGGAUAAG |
| Species: | Anolis carolinensis |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | GUGAGAACUCUGAGGAUAAG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-454-5p | MIMAT0021962 |