Assay Details
Gene Family ID: | MIPF0001000, MIR2109 |
Mature miRNA Sequence: | GGAGGCGUAGAUACUCACACC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Glycine max | gma-miR2109-3p | MIMAT0032101 |
Stem-loop Details
Glycine max
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | gma-miR2109-3p |
|---|---|
| miRBase Accession Number: | MI0010579 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GGAGGCGUAGAUACUCACACC |
| Species: | Glycine max |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0001000, MIR2109 |
Mature miRNA Sequence: | GGAGGCGUAGAUACUCACACC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Glycine max | gma-miR2109-3p | MIMAT0032101 |