Assay Details
Gene Family ID: | MIPF0000183, mir-503 |
Mature miRNA Sequence: | UAGCAGCGGGAACAGUUCUGCAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-503-5p | MIMAT0002874 |
| Gorilla gorilla | ggo-miR-503 | MIMAT0024203 |
| Pan troglodytes | ptr-miR-503 | MIMAT0008176 |
| Pongo pygmaeus | ppy-miR-503 | MIMAT0015926 |
| Rhesus monkey | mml-miR-503-5p | MIMAT0006367 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

Genomic Map