Assay Details
Gene Family ID: | MIPF0000038, mir-1 |
Mature miRNA Sequence: | UGGAAUGUAAAGAAGUAUGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Caenorhabditis elegans | cel-miR-1-3p | MIMAT0000003 |
| Ascaris suum | asu-miR-1-3p | MIMAT0021424 |
| Caenorhabditis brenneri | cbn-miR-1 | MIMAT0029710 |
| Caenorhabditis briggsae | cbr-miR-1 | MIMAT0000465 |
| Chicken | gga-miR-1a-3p | MIMAT0001127 |
| Dog | cfa-miR-1 | MIMAT0006656 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-1 | MIMAT0024078 |
| Heligmosomoides polygyrus | hpo-miR-1-3p | MIMAT0040465 |
| Oryzias latipes | ola-miR-1-3p | MIMAT0022558 |
| Pan paniscus | ppa-miR-1 | MIMAT0002621 |
| Pig | ssc-miR-1 | MIMAT0010187 |
| Python bivittatus | pbv-miR-1a-3p | MIMAT0038813 |
| Xenopus tropicalis | xtr-miR-1a | MIMAT0003551 |
Stem-loop Details
Caenorhabditis elegans
Ascaris suum
Caenorhabditis brenneri
Caenorhabditis briggsae
Chicken
Dog
Gorilla gorilla
Heligmosomoides polygyrus
Oryzias latipes
Pan paniscus
Pig
Python bivittatus
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-1 (v9.2)
Mouse: mmu-miR-1 (v9.2)
Caenorhabditis elegans: cel-miR-1 (v17)
Atlantibacter subterranea: asu-miR-1 (v18)
Gallus gallus: gga-miR-1 (v7.1)
Gallus gallus: gga-miR-1a (v19)
Monodelphis domestica: mdo-miR-1 (v19)
Staphylococcus schleiferi: ssc-miR-1a (v14)
Mouse: mmu-miR-1 (v9.2)
Caenorhabditis elegans: cel-miR-1 (v17)
Atlantibacter subterranea: asu-miR-1 (v18)
Gallus gallus: gga-miR-1 (v7.1)
Gallus gallus: gga-miR-1a (v19)
Monodelphis domestica: mdo-miR-1 (v19)
Staphylococcus schleiferi: ssc-miR-1a (v14)

Genomic Map