Assay Details
Mature miRNA Sequence: | UAGGAUUACAAGUGUCGGCCAC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3159 | MIMAT0015033 |
Stem-loop Details
Human
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | hsa-miR-3159 |
|---|---|
| miRBase Accession Number: | MI0014188 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UAGGAUUACAAGUGUCGGCCAC |
| Species: | Human |
| Product Type: | Ambion® Anti-miR™ miRNA Inhibitor |
Mature miRNA Sequence: | UAGGAUUACAAGUGUCGGCCAC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-3159 | MIMAT0015033 |