Assay Details
Amplicon Length: | 59 |
Chromosome Location: | Chr. 16 - 88468736 - 88468736 [+] on 88468736 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-5189-3p | MIMAT0027088 |
| Human | hsa-miR-5189-5p | MIMAT0021120 |
Stem-loop Details
Human

Genomic Map