Assay Details
Gene Family ID: | MIPF0001753, mir-2062 |
Mature miRNA Sequence: | UGCAACAAUAUUUCAUCAGUGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Branchiostoma belcheri | bbe-miR-2062-5p | MIMAT0031623 |
| Branchiostoma floridae | bfl-miR-2062-5p | MIMAT0009988 |
Stem-loop Details
Branchiostoma belcheri
Branchiostoma floridae
More Information
Related Products
miRBase Alias:
Branchiostoma floridae: bfl-miR-2062 (v18)

Genomic Map