Assay Details
Gene Family ID: | MIPF0000242, mir-425 |
Mature miRNA Sequence: | AAUGACACGAUCACUCCCGCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Ophiophagus hannah | oha-miR-425-5p | MIMAT0036921 |
| Taeniopygia guttata | tgu-miR-425-5p | MIMAT0014470 |
Stem-loop Details
Ophiophagus hannah
Taeniopygia guttata

Genomic Map