Assay Details
Mature miRNA Sequence: | UGGUAACUCCAAACCAUUGCCGG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Acyrthosiphon pisum | api-miR-2765 | MIMAT0014787 |
Stem-loop Details
Acyrthosiphon pisum
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | api-miR-2765 |
|---|---|
| miRBase Accession Number: | MI0014005 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGGUAACUCCAAACCAUUGCCGG |
| Species: | Acyrthosiphon pisum |
| Product Type: | mirVana® miRNA mimic |
Mature miRNA Sequence: | UGGUAACUCCAAACCAUUGCCGG |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Acyrthosiphon pisum | api-miR-2765 | MIMAT0014787 |