Assay Details
Gene Family ID: | MIPF0000242, mir-425 |
Mature miRNA Sequence: | CCACGGGGAUGUCGUGUCUGUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-miR-425-3p | MIMAT0021955 |
Stem-loop Details
Anolis carolinensis
More Information
Related Products
miRBase Alias:
Aeromonas caviae Ae398: aca-miR-425* (v18)

Genomic Map