Assay Details
Gene Family ID: | MIPF0000317, mir-548 |
Mature miRNA Sequence: | AAAAGUAAUUGUGGUUUUUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-548ad-5p | MIMAT0032114 |
Human | hsa-miR-548ae-5p | MIMAT0032115 |
Pongo pygmaeus | ppy-miR-548a | MIMAT0024373 |
Stem-loop Details
Human
Pongo pygmaeus
