Assay Details
Mature miRNA Sequence: | UGGUCGACCAGUUGGAAAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Callithrix jacchus | cja-miR-412 | MIMAT0039499 |
| Rhesus monkey | mml-miR-412-5p | MIMAT0026871 |
Stem-loop Details
Callithrix jacchus
Rhesus monkey

Genomic Map