Assay Details
Gene Family ID: | MIPF0000054, mir-216 |
Mature miRNA Sequence: | UAAAUAUCAGCUGGUAAUUCU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anopheles gambiae | aga-miR-283 | MIMAT0001515 |
| Bombyx mori | bmo-miR-283-5p | MIMAT0004206 |
| Drosophila ananassae | dan-miR-283 | MIMAT0008437 |
| Drosophila erecta | der-miR-283 | MIMAT0008480 |
| Drosophila grimshawi | dgr-miR-283 | MIMAT0008618 |
| Drosophila mojavensis | dmo-miR-283 | MIMAT0008640 |
| View More Rows | ||
| Drosophila persimilis | dpe-miR-283 | MIMAT0008713 |
| Drosophila pseudoobscura | dps-miR-283 | MIMAT0001247 |
| Drosophila sechellia | dse-miR-283 | MIMAT0008838 |
| Drosophila simulans | dsi-miR-283 | MIMAT0008893 |
| Drosophila willistoni | dwi-miR-283 | MIMAT0009014 |
| Drosophila yakuba | dya-miR-283 | MIMAT0009064 |
| Heliconius melpomene | hme-miR-283 | MIMAT0024969 |
Stem-loop Details
Anopheles gambiae
Bombyx mori
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila mojavensis
Drosophila persimilis
Drosophila pseudoobscura
Drosophila sechellia
Drosophila simulans
Drosophila willistoni
Drosophila yakuba
Heliconius melpomene
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-283-5p (v17)
Drosophila melanogaster: dme-miR-283 (v16)
Bombyx mori: bmo-miR-283 (v18)
Drosophila virilis: dvi-miR-283 (v20)
Drosophila melanogaster: dme-miR-283 (v16)
Bombyx mori: bmo-miR-283 (v18)
Drosophila virilis: dvi-miR-283 (v20)

Genomic Map