Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | AAGGAGCUCACAGUCUAUUGAG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-28-5p | MIMAT0000085 |
| Mouse | mmu-miR-28a-5p | MIMAT0000653 |
| Rat | rno-miR-28-5p | MIMAT0000800 |
| Ateles geoffroyi | age-miR-28 | MIMAT0002351 |
| Bovine | bta-miR-28 | MIMAT0009272 |
| Dasypus novemcinctus | dno-miR-28-5p | MIMAT0047547 |
| View More Rows | ||
| Gorilla gorilla | ggo-miR-28 | MIMAT0002354 |
| Guinea pig | cpo-miR-28-5p | MIMAT0046894 |
| Horse | eca-miR-28-5p | MIMAT0013092 |
| Lagothrix lagotricha | lla-miR-28 | MIMAT0002358 |
| Macaca nemestrina | mne-miR-28 | MIMAT0002356 |
| Pan paniscus | ppa-miR-28 | MIMAT0002359 |
| Pan troglodytes | ptr-miR-28 | MIMAT0002353 |
| Pig | ssc-miR-28-5p | MIMAT0002136 |
| Pongo pygmaeus | ppy-miR-28 | MIMAT0002355 |
| Rabbit | ocu-miR-28-5p | MIMAT0048141 |
| Rhesus monkey | mml-miR-28-5p | MIMAT0002352 |
| Saguinus labiatus | sla-miR-28 | MIMAT0002357 |
| Tupaia chinensis | tch-miR-28-5p | MIMAT0036565 |
Stem-loop Details
Human
Mouse
Rat
Ateles geoffroyi
Bovine
Dasypus novemcinctus
Gorilla gorilla
Guinea pig
Horse
Lagothrix lagotricha
Macaca nemestrina
Pan paniscus
Pan troglodytes
Pig
Pongo pygmaeus
Rabbit
Rhesus monkey
Saguinus labiatus
Tupaia chinensis
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-28-5p (v18)
Mouse: mmu-miR-28 (v17)
Rat: rno-miR-28 (v18)
Macaca mulatta: mml-miR-28 (v19)
Mouse: mmu-miR-28 (v17)
Rat: rno-miR-28 (v18)
Macaca mulatta: mml-miR-28 (v19)

Genomic Map