Assay Details
Gene Family ID: | MIPF0000574, mir-1420 |
Mature miRNA Sequence: | AAAUUGCCACCAGGUCAGAUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Ornithorhynchus anatinus | oan-miR-1420g-3p | MIMAT0007319 |
Stem-loop Details
Ornithorhynchus anatinus

More Information
Related Products
miRBase Alias:
Ochrobactrum anthropi: oan-miR-1420g* (v18)
