Assay Details
Gene Family ID: | MIPF0000140, mir-3 |
Mature miRNA Sequence: | UCACUGGGUGAAGUUUGUCGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila grimshawi | dgr-miR-309 | MIMAT0008577 |
| Drosophila mojavensis | dmo-miR-309 | MIMAT0008660 |
| Drosophila virilis | dvi-miR-309-3p | MIMAT0008958 |
Stem-loop Details
Drosophila grimshawi
Drosophila mojavensis
Drosophila virilis

Genomic Map