Assay Details
Gene Family ID: | MIPF0000031, mir-196 |
Mature miRNA Sequence: | UCGACAGCACGACACUGCCUUCA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-196b-3p | MIMAT0017171 |
| Dasypus novemcinctus | dno-miR-196b-3p | MIMAT0047732 |
| Guinea pig | cpo-miR-196b-3p | MIMAT0047083 |
| Rabbit | ocu-miR-196b-3p | MIMAT0048316 |
Stem-loop Details
Rat
Dasypus novemcinctus
Guinea pig
Rabbit

Genomic Map