Assay Details
Amplicon Length: | 67 |
Chromosome Location: | Chr. 2 - 10466545 - 10466545 [+] on 10466545 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-466f-3p | MIMAT0004882 |
| Mouse | mmu-miR-466f-5p | MIMAT0004881 |
| Mouse | mmu-miR-669f-3p | MIMAT0005839 |
| Mouse | mmu-miR-669f-5p | MIMAT0017327 |
Stem-loop Details
Mouse

Genomic Map