Assay Details
Amplicon Length: | 95 |
Chromosome Location: | Chr. X - 72991927 - 72991927 [-] on 72991927 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-2137 | MIMAT0011213 |
Stem-loop Details
Mouse
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 95 |
Chromosome Location: | Chr. X - 72991927 - 72991927 [-] on 72991927 Build GRCm38 |
Species: | Mouse |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-2137 | MIMAT0011213 |