Assay Details
Gene Family ID: | MIPF0000192, mir-412 |
Mature miRNA Sequence: | ACUUCACCUGGUCCACUAGCCGU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-412-3p | MIMAT0002170 |
Pan troglodytes | ptr-miR-412 | MIMAT0008139 |
Pongo pygmaeus | ppy-miR-412 | MIMAT0015879 |
Rhesus monkey | mml-miR-412-3p | MIMAT0006318 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-412 (v19)
Mouse: mmu-miR-412 (v9.2)
Rat: rno-miR-412 (v15)
Macaca mulatta: mml-miR-412 (v19)
Mouse: mmu-miR-412 (v9.2)
Rat: rno-miR-412 (v15)
Macaca mulatta: mml-miR-412 (v19)
