Assay Details
Gene Family ID: | MIPF0000033, mir-10 |
Mature miRNA Sequence: | CAAGCUCGCUUCUAUGGGUCUG |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-99a-3p | MIMAT0004511 |
| Alligator mississippiensis | ami-miR-99a-3p | MIMAT0038111 |
| Chrysemys picta | cpi-miR-99a-3p | MIMAT0037711 |
| Columba livia | cli-miR-99-3p | MIMAT0038479 |
| Cricetulus griseus | cgr-miR-99a-3p | MIMAT0024017 |
| Dasypus novemcinctus | dno-miR-99a-3p | MIMAT0047595 |
| View More Rows | ||
| Guinea pig | cpo-miR-99a-3p | MIMAT0046942 |
| Rabbit | ocu-miR-99a-3p | MIMAT0048183 |
Stem-loop Details
Human
Alligator mississippiensis
Chrysemys picta
Columba livia
Cricetulus griseus
Dasypus novemcinctus
Guinea pig
Rabbit

Genomic Map