Assay Details
Gene Family ID: | MIPF0000022, mir-7 |
Mature miRNA Sequence: | CAACAAGUCCCAGUCUGCCACA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-7a-2-3p | MIMAT0017070 |
| Rat | rno-miR-7a-2-3p | MIMAT0017091 |
| Pteropus alecto | pal-miR-7a-3p | MIMAT0040113 |
Stem-loop Details
Mouse
Rat
Pteropus alecto
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-7a-2* (v17)
Rat: rno-miR-7a-2* (v18)
Rat: rno-miR-7a-2* (v18)

Genomic Map