Assay Details
Amplicon Length: | 77 |
Chromosome Location: | Chr. 2 - 225873804 - 225873804 [+] on 225873804 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-760-3p | MIMAT0005337 |
| Rat | rno-miR-760-5p | MIMAT0005336 |
Stem-loop Details
Rat

Genomic Map