Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 13 - 80125735 - 80125735 [-] on 80125735 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-199a-3p | MIMAT0004738 |
| Rat | rno-miR-199a-5p | MIMAT0000872 |
Stem-loop Details
Rat

Genomic Map