Assay Details
Amplicon Length: | 127 |
Chromosome Location: | Chr. 1 - 122373623 - 122373623 [-] on 122373623 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-344a-5p | MIMAT0004654 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 127 |
Chromosome Location: | Chr. 1 - 122373623 - 122373623 [-] on 122373623 Build Rnor_6.0 |
Species: | Rat |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-344a-5p | MIMAT0004654 |