Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 20 - 3319121 - 3319121 [+] on 3319121 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-877 | MIMAT0005285 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 92 |
Chromosome Location: | Chr. 20 - 3319121 - 3319121 [+] on 3319121 Build Rnor_6.0 |
Species: | Rat |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-877 | MIMAT0005285 |