Assay Details
Amplicon Length: | 94 |
Chromosome Location: | Chr. 6 - 133879016 - 133879016 [-] on 133879016 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-544-5p | MIMAT0017359 |
| Rat | rno-miR-544-3p | MIMAT0012831 |
| Rat | rno-miR-6331 | MIMAT0025070 |
Stem-loop Details
Rat

Genomic Map