Assay Details
Gene Family ID: | MIPF0000313, mir-306 |
Mature miRNA Sequence: | UCAGGUACUUAGUGACUCUCAA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Drosophila melanogaster | dme-miR-306-5p | MIMAT0000393 | |
| Drosophila ananassae | dan-miR-306 | MIMAT0008450 | |
| Drosophila erecta | der-miR-306-5p | MIMAT0008494 | |
| Drosophila grimshawi | dgr-miR-306 | MIMAT0008592 | |
| Drosophila mojavensis | dmo-miR-306-5p | MIMAT0008648 | |
| Drosophila persimilis | dpe-miR-306 | MIMAT0008759 | |
| View More Rows | |||
| Drosophila pseudoobscura | dps-miR-306 | MIMAT0001256 | |
| Drosophila sechellia | dse-miR-306-5p | MIMAT0008804 | |
| Drosophila willistoni | dwi-miR-306 | MIMAT0008986 | |
| Drosophila yakuba | dya-miR-306-5p | MIMAT0009068 | |
Stem-loop Details
Drosophila melanogaster
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila mojavensis
Drosophila persimilis
Drosophila pseudoobscura
Drosophila sechellia
Drosophila willistoni
Drosophila yakuba
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-306 (v16)
Drosophila erecta: der-miR-306 (v18)
Drosophila mojavensis: dmo-miR-306 (v18)
Drosophila sechellia: dse-miR-306 (v18)
Drosophila virilis: dvi-miR-306-5p (v20)
Drosophila virilis: dvi-miR-306 (v18)
Drosophila yakuba: dya-miR-306 (v18)
Drosophila erecta: der-miR-306 (v18)
Drosophila mojavensis: dmo-miR-306 (v18)
Drosophila sechellia: dse-miR-306 (v18)
Drosophila virilis: dvi-miR-306-5p (v20)
Drosophila virilis: dvi-miR-306 (v18)
Drosophila yakuba: dya-miR-306 (v18)

Genomic Map