Assay Details
Gene Family ID: | MIPF0000566, mir-310 |
Mature miRNA Sequence: | UAUUGCACACUUCCCGGCCUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Drosophila melanogaster | dme-miR-310-3p | MIMAT0000402 |
| Drosophila sechellia | dse-miR-310-3p | MIMAT0008779 |
| Drosophila simulans | dsi-miR-310-3p | MIMAT0008841 |
Stem-loop Details
Drosophila melanogaster
Drosophila sechellia
Drosophila simulans
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-310 (v16)
Drosophila sechellia: dse-miR-310 (v21)
Drosophila simulans: dsi-miR-310 (v21)
Drosophila sechellia: dse-miR-310 (v21)
Drosophila simulans: dsi-miR-310 (v21)

Genomic Map