Assay Details
Gene Family ID: | MIPF0000141, mir-315 |
Mature miRNA Sequence: | UUUUGAUUGUUGCUCAGAAAGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Drosophila melanogaster | dme-miR-315-5p | MIMAT0000407 | |
| Aedes aegypti | aae-miR-315-5p | MIMAT0014262 | |
| Anopheles gambiae | aga-miR-315 | MIMAT0001521 | |
| Apis mellifera | ame-miR-315-5p | MIMAT0001487 | |
| Culex quinquefasciatus | cqu-miR-315 | MIMAT0014365 | |
| Daphnia pulex | dpu-miR-315 | MIMAT0012660 | |
| View More Rows | |||
| Drosophila ananassae | dan-miR-315 | MIMAT0008459 | |
| Drosophila erecta | der-miR-315 | MIMAT0008539 | |
| Drosophila grimshawi | dgr-miR-315 | MIMAT0008604 | |
| Drosophila mojavensis | dmo-miR-315 | MIMAT0008687 | |
| Drosophila persimilis | dpe-miR-315 | MIMAT0008730 | |
| Drosophila pseudoobscura | dps-miR-315 | MIMAT0001261 | |
| Drosophila sechellia | dse-miR-315 | MIMAT0008815 | |
| Drosophila simulans | dsi-miR-315 | MIMAT0008870 | |
| Drosophila willistoni | dwi-miR-315 | MIMAT0009036 | |
| Drosophila yakuba | dya-miR-315 | MIMAT0009084 | |
| Nasonia longicornis | nlo-miR-315 | MIMAT0018433 | |
| Nasonia vitripennis | nvi-miR-315 | MIMAT0015689 | |
| Tribolium castaneum | tca-miR-315 | MIMAT0010096 | |
Stem-loop Details
Drosophila melanogaster
Aedes aegypti
Anopheles gambiae
Apis mellifera
Culex quinquefasciatus
Daphnia pulex
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila mojavensis
Drosophila persimilis
Drosophila pseudoobscura
Drosophila sechellia
Drosophila simulans
Drosophila willistoni
Drosophila yakuba
Nasonia longicornis
Nasonia vitripennis
Tribolium castaneum
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-315 (v16)
Achromobacter aegrifaciens: aae-miR-315 (v18)
Anaerococcus mediterraneensis: ame-miR-315 (v21)
Drosophila virilis: dvi-miR-315a (v20)
Achromobacter aegrifaciens: aae-miR-315 (v18)
Anaerococcus mediterraneensis: ame-miR-315 (v21)
Drosophila virilis: dvi-miR-315a (v20)

Genomic Map