Assay Details
Gene Family ID: | MIPF0000254, mir-316 |
Mature miRNA Sequence: | UGUCUUUUUCCGCUUACUGGCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Drosophila melanogaster | dme-miR-316-5p | MIMAT0000408 | |
| Drosophila ananassae | dan-miR-316 | MIMAT0008449 | |
| Drosophila erecta | der-miR-316 | MIMAT0008496 | |
| Drosophila grimshawi | dgr-miR-316 | MIMAT0008591 | |
| Drosophila persimilis | dpe-miR-316 | MIMAT0008764 | |
| Drosophila pseudoobscura | dps-miR-316 | MIMAT0001262 | |
| View More Rows | |||
| Drosophila sechellia | dse-miR-316 | MIMAT0008818 | |
| Drosophila simulans | dsi-miR-316 | MIMAT0008869 | |
| Drosophila willistoni | dwi-miR-316 | MIMAT0009000 | |
| Drosophila yakuba | dya-miR-316 | MIMAT0009080 | |
Stem-loop Details
Drosophila melanogaster
Drosophila ananassae
Drosophila erecta
Drosophila grimshawi
Drosophila persimilis
Drosophila pseudoobscura
Drosophila sechellia
Drosophila simulans
Drosophila willistoni
Drosophila yakuba
More Information
Related Products
miRBase Alias:
Drosophila melanogaster: dme-miR-316 (v16)
Drosophila virilis: dvi-miR-316 (v20)
Drosophila virilis: dvi-miR-316 (v20)

Genomic Map