Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | AGAGGUAGUAGGUUGCAUAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Anolis carolinensis | aca-let-7d-5p | MIMAT0021699 |
| Monodelphis domestica | mdo-let-7d | MIMAT0004172 |
Stem-loop Details
Anolis carolinensis
Monodelphis domestica
More Information
Related Products
miRBase Alias:
Human: hsa-let-7d (v9.2)
Mouse: mmu-let-7d (v9.2)
Rat: rno-let-7d (v9.2)
Aeromonas caviae Ae398: aca-let-7d (v18)
Mouse: mmu-let-7d (v9.2)
Rat: rno-let-7d (v9.2)
Aeromonas caviae Ae398: aca-let-7d (v18)

Genomic Map