Assay Details
Gene Family ID: | MIPF0000002, let-7 |
Mature miRNA Sequence: | UGAGGUAGGAGGUUGUAUAGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Bovine | bta-let-7e | MIMAT0004333 |
| Gorilla gorilla | ggo-let-7e | MIMAT0024079 |
Stem-loop Details
Bovine
Gorilla gorilla
More Information
Related Products
miRBase Alias:
Human: hsa-let-7e (v9.2)
Mouse: mmu-let-7e (v9.2)
Rat: rno-let-7e (v9.2)
Mouse: mmu-let-7e (v9.2)
Rat: rno-let-7e (v9.2)

Genomic Map