Assay Details
Gene Family ID: | MIPF0000078, mir-187 |
Mature miRNA Sequence: | UCGUGUCUUGUGUUGCAGCCG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Cricetulus griseus | cgr-miR-187 | MIMAT0023815 |
| Gorilla gorilla | ggo-miR-187 | MIMAT0002515 |
| Macaca nemestrina | mne-miR-187 | MIMAT0002517 |
| Pan paniscus | ppa-miR-187 | MIMAT0002518 |
| Pongo pygmaeus | ppy-miR-187 | MIMAT0002516 |
Stem-loop Details
Cricetulus griseus
Gorilla gorilla
Macaca nemestrina
Pan paniscus
Pongo pygmaeus
More Information
Related Products
miRBase Alias:
Human: hsa-miR-187 (v9.2)
Rat: rno-miR-187 (v9.2)
Rat: rno-miR-187 (v9.2)

Genomic Map